Learn More
Justrite EN Cabinet Mover
Protect workers, reduce fire risks, and improve productivity by storing flammable fuels and chemicals in code-compliant safety cabinets. Designed to meet OSHA and NFPA standards.
Brand: Justrite JCB22638BL
Additional Details : Weight : 8.20000kg
Description
- Protect workers, reduce fire risks, and improve productivity by storing flammable fuels and chemicals in code-compliant safety cabinets
- Designed to meet OSHA and NFPA standards, Sure-Grip™ EX cabinets are constructed of sturdy 18 gauge (1 mm) thick double-wall, welded steel with 1 1/2 in. (38 mm) of insulating air space for fire resistance
- High-performance, self-latching doors close easily and securely for maximum protection under fire conditions
- Fail-safe closing mechanism ensures three-point stainless steel bullet latching system works every time
Specifications
Black | |
Cabinet Mover | |
60 cm | |
14 cm |