missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Enolase 2/Neuron-specific Enolase Antibody (NSE-P2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30895-200ug
This item is not returnable.
View return policy
Description
Enolase 2/Neuron-specific Enolase Monoclonal antibody specifically detects Enolase 2/Neuron-specific Enolase in Chicken,Human,Mouse samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Enolase 2/Neuron-specific Enolase | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Chicken, Human, Mouse | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| NSE-P2 | |
| Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL | |
| 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the human Enolase 2/Neuron-specific Enolase protein and mouse myeloma Ag8563 cells. Sequence common in human, mouse and chicken. | |
| 200 μg | |
| Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 2026 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction