missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENOX2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10423-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a tumor-specific member of the ECTO-NOX family of genes that encode cell surface NADH oxidases. The encoded protein has two enzymatic activities: catalysis of hydroquinone or NADH oxidation, and protein disulfide interchange. The protein also displays prion-like properties. Alternative splicing results in multiple transcript variants.Specifications
| ENOX2 | |
| Polyclonal | |
| Unconjugated | |
| ENOX2 | |
| APK1, APK1 antigen, COVA1, ENOX2, tNOX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10495, 209224 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16206, Q8R0Z2 | |
| ENOX2 | |
| ENOX2 Fusion Protein Ag0674 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction