missing translation for 'onlineSavingsMsg'
Learn More

ent-Paroxetine Hydrochloride, TRC

Product Code. 30423464
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423464 1 g -
30411031 25 mg -
30418761 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423464 Supplier Toronto Research Chemicals Supplier No. TRCP2057551G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130855-30-0
Molecular Formula C19 H20 F N O3 . Cl H
Molecular Weight (g/mol) 365.83
Synonym (3R,4S)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, (3R,4S)- (9CI), Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-, hydrochloride, (3R-trans)-, Paroxetine Related Compound C (USP), Paroxetine Imp. D (EP), (+)-trans-Paroxetine Hydrochloride
IUPAC Name (3R,4S)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)piperidine;hydrochloride
SMILES Cl.Fc1ccc(cc1)[C@H]2CCNC[C@@H]2COc3ccc4OCOc4c3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H20FNO3.ClH/c20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18;/h1-6,9,14,17,21H,7-8,10-12H2;1H/t14-,17-;/m1./s1
Formula Weight 365.1194
Percent Purity >95%
Chemical Name or Material (+)-trans-paroxetine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.