missing translation for 'onlineSavingsMsg'
Learn More

ent-Ritlecitinib, TRC

Product Code. 30416003
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416003 50 mg 50mg
30413249 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30416003 Supplier Toronto Research Chemicals Supplier No. TRCR29982050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ent-Ritlecitinib
CAS 1792180-79-0
Molecular Formula C15H19N5O
Quantity 50 mg
Synonym 1-[(2R,5S)-2-Methyl-5-(7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)-1-piperidinyl]-2-propen-1-one, (2R,5S)-Ritlecitinib
InChI Formula InChI=1S/C15H19N5O/c1-3-13(21)20-8-11(5-4-10(20)2)19-15-12-6-7-16-14(12)17-9-18-15/h3,6-7,9-11H,1,4-5,8H2,2H3,(H2,16,17,18,19)/t10-,11+/m1/s1
SMILES N(C1=C2C(=NC=N1)NC=C2)[C@@H]3CN(C(C=C)=O)[C@H](C)CC3
IUPAC Name 1-((2R,5S)-5-((7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)-2-methylpiperidin-1-yl)prop-2-en-1-one
Molecular Weight (g/mol) 285.34
Formula Weight 285.159
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.