missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Eotaxin Monoclonal Antibody (43911)

Mouse Monoclonal Antibody
458.00€ - 688.00€
Specifications
| Antigen | Eotaxin |
|---|---|
| Clone | 43911 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin), Neutralization, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15523432
|
Invitrogen™
MA523739 |
500 μg |
688.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15543522
![]() |
Invitrogen™
MA523831 |
100 μg |
458.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.Specifications
| Eotaxin | |
| 0.5 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 5% trehalose and No Preservative | |
| P51671 | |
| 6356 | |
| E. coli-derived recombinant human CCL11/Eotaxin Gly24-Pro97. | |
| Antibody |
| 43911 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| CCL11 | |
| C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11 | |
| CCL11 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title