missing translation for 'onlineSavingsMsg'
Learn More

EPA 525.2, HJ 867-2017 Labelled PAH Mixture 158 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30327221
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30327221

Brand: Dr. Ehrenstorfer DREA50000158DI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acenaphthene D10∣Chrysene D12∣Perylene D12∣Phenanthrene D10
CAS 15067-26-2, 1719-03-5, 1520-96-3, 1517-22-2
Analyte or Component Names Acenaphthene D10∣Chrysene D12∣Perylene D12∣Phenanthrene D10
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Acenaphthene D10 2000 μg/mL∣Chrysene D12 2000 μg/mL∣Perylene D12 2000 μg/mL∣Phenanthrene D10 2000 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.