missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 515.2 Methyl Derivatives Mixture 403 100-500 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30325699
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325699 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325699 Supplier Dr. Ehrenstorfer Supplier No. DREA50000403ME
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,5-T-methyl ester∣3,5-Dichlorobenzoic acid methyl ester∣Chlorthal-dimethyl∣Dichlorprop-methyl ester∣Dinoseb methyl ether∣Pentachloroanisole
CAS 1928-37-6, 2905-67-1, 1861-32-1, 23844-57-7, 6099-79-2, 1825-21-4
Analyte or Component Names 2,4,5-T-methyl ester∣3,5-Dichlorobenzoic acid methyl ester∣Chlorthal-dimethyl∣Dichlorprop-methyl ester∣Dinoseb methyl ether∣Pentachloroanisole
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4,5-T-methyl ester 100 μg/mL∣Chlorthal-dimethyl 100 μg/mL∣Dichlorprop-methyl ester 100 μg/mL∣Pentachloroanisole 100 μg/mL∣Dinoseb methyl ether 200 μg/mL∣3,5-Dichlorobenzoic acid methyl ester 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.