missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 525.2 Organonitrogen Pesticide Mixture 500 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30321159
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321159 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321159 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000339AC
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alachlor∣Ametryn∣Atraton∣Atrazine∣Bromacil∣Butachlor∣Butylate∣Chlorpropham∣Cyanazine∣Cycloate∣Diphenamid∣EPTC∣Etridiazole∣Fenarimol∣Fluridone∣Hexazinone∣Metolachlor∣MGK 264∣Molinate∣Napropamide∣Norflurazon∣Pebulate∣Prometon∣Prometryn∣Propachlor∣Propazine∣Propyzamide∣Simazine∣Simetryn∣Tebuthiuron∣Terbacil∣Terbutryn∣Triadimefon∣Tricyclazole∣Trifluralin∣Vernolate
CAS 15972-60-8, 834-12-8, 1610-17-9, 1912-24-9, 314-40-9, 23184-66-9
Analyte or Component Names Alachlor∣Ametryn∣Atraton∣Atrazine∣Bromacil∣Butachlor∣Butylate∣Chlorpropham∣Cyanazine∣Cycloate∣Diphenamid∣EPTC∣Etridiazole∣Fenarimol∣Fluridone∣Hexazinone∣Metolachlor∣MGK 264∣Molinate∣Napropamide∣Norflurazon∣Pebulate∣Prometon∣Prometryn∣Propachlor∣Propazine∣Propyzamide∣Simazine∣Simetryn∣Tebuthiuron∣Terbacil∣Terbutryn∣Triadimefon∣Tricyclazole∣Trifluralin∣Vernolate
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Alachlor 500 μg/mL∣Ametryn 500 μg/mL∣Atraton 500 μg/mL∣Atrazine 500 μg/mL∣Bromacil 500 μg/mL∣Butachlor 500 μg/mL∣Butylate 500 μg/mL∣Chlorpropham 500 μg/mL∣Cyanazine 500 μg/mL∣Cycloate 500 μg/mL∣Diphenamid 500 μg/mL∣EPTC 500 μg/mL∣Etridiazole 500 μg/mL∣Fenarimol 500 μg/mL∣Fluridone 500 μg/mL∣Hexazinone 500 μg/mL∣Metolachlor 500 μg/mL∣MGK 264 500 μg/mL∣Molinate 500 μg/mL∣Napropamide 500 μg/mL∣Norflurazon 500 μg/mL∣Pebulate 500 μg/mL∣Prometon 500 μg/mL∣Prometryn 500 μg/mL∣Propachlor 500 μg/mL∣Propazine 500 μg/mL∣Propyzamide 500 μg/mL∣Simazine 500 μg/mL∣Simetryn 500 μg/mL∣Tebuthiuron 500 μg/mL∣Terbacil 500 μg/mL∣Terbutryn 500 μg/mL∣Triadimefon 500 μg/mL∣Tricyclazole 500 μg/mL∣Trifluralin 500 μg/mL∣Vernolate 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.