Learn More
EPA Method 525.3 PCB Mixture 500 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | PCB 3 (4-Chlorobiphenyl)∣PCB No. 1∣PCB No. 110∣PCB No. 118∣PCB No. 138∣PCB No. 149∣PCB No. 153∣PCB No. 18∣PCB No. 180∣PCB No. 28∣PCB No. 44∣PCB No. 52∣PCB No. 70∣PCB No. 8 |
| CAS | 2051-62-9, 2051-60-7, 38380-03-9, 31508-00-6, 35065-28-2, 38380-04-0 |
| Analyte or Component Names | PCB 3 (4-Chlorobiphenyl)∣PCB No. 1∣PCB No. 110∣PCB No. 118∣PCB No. 138∣PCB No. 149∣PCB No. 153∣PCB No. 18∣PCB No. 180∣PCB No. 28∣PCB No. 44∣PCB No. 52∣PCB No. 70∣PCB No. 8 |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | PCB 3 (4-Chlorobiphenyl) 500 μg/mL∣PCB No. 1 500 μg/mL∣PCB No. 110 500 μg/mL∣PCB No. 118 500 μg/mL∣PCB No. 138 500 μg/mL∣PCB No. 149 500 μg/mL∣PCB No. 153 500 μg/mL∣PCB No. 18 500 μg/mL∣PCB No. 180 500 μg/mL∣PCB No. 28 500 μg/mL∣PCB No. 44 500 μg/mL∣PCB No. 52 500 μg/mL∣PCB No. 70 500 μg/mL∣PCB No. 8 500 μg/mL |