missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 6020 Ca, Fe, K, Mg, Na @ 2000 mg/l, 100mls. Part of Kit # 4600432 , VHG™

Product Code. 17988699
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988699 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17988699 Supplier VHG Labs Supplier No. VHG1600601
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

EPA Method 6020 Ca, Fe, K, Mg, Na @ 2000 mg/l, 100mls. Part of Kit # 4600432
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium/Iron/Magnesium/Potassium/Sodium
CAS 7440-70-2, 7439-89-6, 7439-95-4, 9-7-7440, 7440-23-5
Methods ICP/ICP-MS
Analyte or Component Names Calcium/Iron/Magnesium/Potassium/Sodium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Calcium 2000 mg/L/Iron 2000 mg/L/Magnesium 2000 mg/L/Potassium 2000 mg/L/Sodium 2000 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.