missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 625 Mixture 247 1000-2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30320836
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30320836 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30320836

Brand: Dr. Ehrenstorfer DREA50000247DI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,4,5-Tetrachlorophenol∣2,3,4,6-Tetrachlorophenol∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dinitrotoluene∣2,6-Dichlorophenol∣2-Chlorophenol∣2-Methylphenol∣3,4-Dichlorophenol∣3-Chlorophenol∣3-Methylphenol (m-Cresol)∣4-Chlorophenol∣4-Methylphenol (p-Cresol)∣Carbazole∣Fluoranthene∣n-Decane∣Nitrobenzene∣n-Octadecane∣Pentachlorophenol∣Phenol∣Phthalic acid, bis-2-ethylhexyl ester
CAS 4901-51-3, 58-90-2, 95-95-4, 88-06-2, 120-83-2, 121-14-2
Analyte or Component Names 2,3,4,5-Tetrachlorophenol∣2,3,4,6-Tetrachlorophenol∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dinitrotoluene∣2,6-Dichlorophenol∣2-Chlorophenol∣2-Methylphenol∣3,4-Dichlorophenol∣3-Chlorophenol∣3-Methylphenol (m-Cresol)∣4-Chlorophenol∣4-Methylphenol (p-Cresol)∣Carbazole∣Fluoranthene∣n-Decane∣Nitrobenzene∣n-Octadecane∣Pentachlorophenol∣Phenol∣Phthalic acid, bis-2-ethylhexyl ester
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 3-Methylphenol (m-Cresol) 1000 μg/mL∣4-Methylphenol (p-Cresol) 1000 μg/mL∣2,3,4,5-Tetrachlorophenol 2000 μg/mL∣2,3,4,6-Tetrachlorophenol 2000 μg/mL∣2,4,5-Trichlorophenol 2000 μg/mL∣2,4,6-Trichlorophenol 2000 μg/mL∣2,4-Dichlorophenol 2000 μg/mL∣2,4-Dinitrotoluene 2000 μg/mL∣2,6-Dichlorophenol 2000 μg/mL∣2-Chlorophenol 2000 μg/mL∣2-Methylphenol 2000 μg/mL∣3,4-Dichlorophenol 2000 μg/mL∣3-Chlorophenol 2000 μg/mL∣4-Chlorophenol 2000 μg/mL∣Carbazole 2000 μg/mL∣Fluoranthene 2000 μg/mL∣n-Decane 2000 μg/mL∣Nitrobenzene 2000 μg/mL∣n-Octadecane 2000 μg/mL∣Pentachlorophenol 2000 μg/mL∣Phenol 2000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.