missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8260 Internal Standard Mixture 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1,4-Dichlorobenzene-D4∣Chlorobenzene D5∣Fluorobenzene |
| CAS | 3855-82-1, 3114-55-4, 462-06-6 |
| Analyte or Component Names | 1,4-Dichlorobenzene-D4∣Chlorobenzene D5∣Fluorobenzene |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Industry Type | Food Testing |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,4-Dichlorobenzene-D4 2000 μg/mL∣Chlorobenzene D5 2000 μg/mL∣Fluorobenzene 2000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?