missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 Acid Surrogate Mixture 10000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30328266
Change view
Click to view available options
Quantity:
5 x 5 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328266 5 x 5 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30328266 Supplier Dr. Ehrenstorfer Supplier No. DRESY09000025ME
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Tribromophenol∣2-Fluorophenol∣Phenol D6
CAS 118-79-6, 367-12-4, 13127-88-3
Analyte or Component Names 2,4,6-Tribromophenol∣2-Fluorophenol∣Phenol D6
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Industry Type Food Testing
Quantity 5 x 5 mL
Concentration or Composition (by Analyte or Components) 2,4,6-Tribromophenol 10000 μg/mL∣2-Fluorophenol 10000 μg/mL∣Phenol D6 10000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.