missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 8270 BN Mixture 207 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣Azobenzene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Isophorone∣Nitrobenzene |
| CAS | 120-82-1, 95-50-1, 541-73-1, 106-46-7, 121-14-2, 606-20-2 |
| Analyte or Component Names | 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣Azobenzene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Isophorone∣Nitrobenzene |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,2,4-Trichlorobenzene 2000 μg/mL∣1,2-Dichlorobenzene 2000 μg/mL∣1,3-Dichlorobenzene 2000 μg/mL∣1,4-Dichlorobenzene 2000 μg/mL∣2,4-Dinitrotoluene 2000 μg/mL∣2,6-Dinitrotoluene 2000 μg/mL∣2-Chloronaphthalene 2000 μg/mL∣Azobenzene 2000 μg/mL∣Hexachlorobenzene 2000 μg/mL∣Hexachlorobutadiene 2000 μg/mL∣Hexachlorocyclopentadiene 2000 μg/mL∣Hexachloroethane 2000 μg/mL∣Isophorone 2000 μg/mL∣Nitrobenzene 2000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?