Learn More
EPA Method 8270 Internal Standard Mixture 4000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Product Code. 30323752
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323752 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323752 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000429DI
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,4-Dichlorobenzene-D4∣1,4-Dioxane D8∣Acenaphthene D10∣Chrysene D12∣Naphthalene D8∣Perylene D12∣Phenanthrene D10
CAS 3855-82-1, 17647-74-4, 15067-26-2, 1719-03-5, 1146-65-2, 1520-96-3
Analyte or Component Names 1,4-Dichlorobenzene-D4∣1,4-Dioxane D8∣Acenaphthene D10∣Chrysene D12∣Naphthalene D8∣Perylene D12∣Phenanthrene D10
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) 1,4-Dioxane D8 1000 μg/mL∣1,4-Dichlorobenzene-D4 4000 μg/mL∣Acenaphthene D10 4000 μg/mL∣Chrysene D12 4000 μg/mL∣Naphthalene D8 4000 μg/mL∣Perylene D12 4000 μg/mL∣Phenanthrene D10 4000 μg/mL