Learn More
EPA Method 8270 Organochlorine Pesticides Mixture 1000 μg/mL in Methyl tert-butyl ether, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hydroquinone∣Methoxychlor∣Trans-Chlordane (Gamma Isomer) |
| CAS | 72-54-8, 72-55-9, 50-29-3, 309-00-2, 959-98-8, 319-84-6 |
| Analyte or Component Names | 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hydroquinone∣Methoxychlor∣Trans-Chlordane (Gamma Isomer) |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | MTBE |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 4,4'-DDD 1000 μg/mL∣4,4'-DDE 1000 μg/mL∣4,4'-DDT 1000 μg/mL∣Aldrin 1000 μg/mL∣alpha-Endosulfan 1000 μg/mL∣alpha-HCH 1000 μg/mL∣beta-Endosulfan 1000 μg/mL∣beta-HCH 1000 μg/mL∣Cis-Chlordane (Alpha Isomer) 1000 μg/mL∣delta-HCH 1000 μg/mL∣Dieldrin 1000 μg/mL∣Endosulfan-sulfate 1000 μg/mL∣Endrin 1000 μg/mL∣Endrin-aldehyde 1000 μg/mL∣Endrin-ketone 1000 μg/mL∣gamma-HCH 1000 μg/mL∣Heptachlor 1000 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 1000 μg/mL∣Hydroquinone 1000 μg/mL∣Methoxychlor 1000 μg/mL∣Trans-Chlordane (Gamma Isomer) 1000 μg/mL |