missing translation for 'onlineSavingsMsg'
Learn More

EPA VOC Additional Compounds Mixture 2000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30317398
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317398 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317398 Supplier Dr. Ehrenstorfer Supplier No. DREYA09000012ME
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Butanone∣2-Chloroethyl-vinylether∣2-Hexanone∣4-Methyl-2-pentanone (MIBK)∣Acetone∣Carbon disulfide∣Methyl iodide∣Vinylacetate
CAS 78-93-3, 110-75-8, 591-78-6, 108-10-1, 67-64-1, 75-15-0
Analyte or Component Names 2-Butanone∣2-Chloroethyl-vinylether∣2-Hexanone∣4-Methyl-2-pentanone (MIBK)∣Acetone∣Carbon disulfide∣Methyl iodide∣Vinylacetate
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Industry Type Food Testing
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2-Butanone 2000 μg/mL∣2-Chloroethyl-vinylether 2000 μg/mL∣2-Hexanone 2000 μg/mL∣4-Methyl-2-pentanone (MIBK) 2000 μg/mL∣Acetone 2000 μg/mL∣Carbon disulfide 2000 μg/mL∣Methyl iodide 2000 μg/mL∣Vinylacetate 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.