missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio Epcam / CD326 Rabbit, Clone: EGP40/1555R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 4072-RBM15-P1
Additional Details : Weight : 0.01000kg
Specifications
Epcam / CD326 | |
Cell surface | |
EGP40/1555R | |
Unconjugated | |
NP_002345 | |
Rabbit | |
40-43kDa | |
Protein A/G Purified | |
RUO | |
4072 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
EPCAM | |
A synthetic peptide (around aa 20-60) from the N-terminus of human EPCAM protein | |
HT29 cells or breast tumor | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |