Learn More
EpCAM/TROP1 Antibody (CL6259) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76485IR
Product Code. 30292644
  • 737.00€ / 0.10mL
Description
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EpCAM/TROP1
Monoclonal
DyLight 755
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
This antibody was developed using a synthetic peptide derived from P16422, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cancer Stem Cells, Tumor Biomarkers
4072
Store at 4⪚C in the dark.
IgG3
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL6259
50mM Sodium Borate
Mouse
Protein A purified