missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPCR Antibody (RCR-252) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB600-963
This item is not returnable.
View return policy
Description
EPCR Monoclonal specifically detects EPCR in Human samples. It is validated for Flow Cytometry, Microarray, Block/Neutralize.
Specifications
| EPCR | |
| Monoclonal | |
| 2.4 mg/mL | |
| Flow Cytometry 5-20 ug/ml, Blocking/Neutralizing 50-200 molar excess, Microarray | |
| PROCR | |
| Myeloma cells and cells isolated from the superficial inguinal lymph nodes from Wistar rats immunized with the human EPCR-positive RE-1 cells. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Blocking Assay | |
| RCR-252 | |
| Unconjugated | |
| APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial | |
| Rat | |
| 49 kDa | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 10544 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction