Learn More
BRAND™ EPDM Rubber Conical Gaskets
Product Code. 10614552 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric) Outside:
11 mm
16 mm
20 mm
22 to 84 mm
25 mm
32 mm
43 mm
71 mm
89 mm
Unit Size:
Pack of 10
Pack of 8
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside unitSize
10614552 22 to 84 mm Pack of 8
10548781 11 mm Pack of 10
10689071 16 mm Pack of 10
10518971 20 mm Pack of 10
10325571 43 mm Pack of 10
10277971 71 mm Pack of 10
10101971 25 mm Pack of 10
10049900 32 mm Pack of 10
10599531 89 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10614552 Supplier BRAND™ Supplier No. 462500
Only null left
Add to basket
Add to basket
Place between filter funnel and filter flask for a secure fit.
  • Rubber (EPDM) construction
  • Conical shape
  • Provides a secure fit between filter funnel and filter flask
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inside 22 to 84 mm dia.
For Use With (Application) Used to Place between Filter Funnel and Filter Flask
Quantity 8
Size (Metric) 22 to 84 mm
Diameter (Metric) Outside 22 to 84 mm
Material EPDM
Type Gasket Set