Learn More
EPHX2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10833-1-AP-20UL
 View more versions of this product
Product Code. 16837483
  • 145.00€ / 20µL
Description
This gene encodes a member of the epoxide hydrolase family. The protein, found in both the cytosol and peroxisomes, binds to specific epoxides and converts them to the corresponding dihydrodiols. Mutations in this gene have been associated with familial hypercholesterolemia. Alternatively spliced transcript variants have been described.
Specifications
Specifications
EPHX2
Polyclonal
Unconjugated
EPHX2
CEH, Cytosolic epoxide hydrolase, EPHX2, Epoxide hydratase, Epoxide hydrolase 2, SEH, Soluble epoxide hydrolase
Rabbit
Antigen Affinity Chromatography
RUO
2053
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P34913
EPHX2
EPHX2 Fusion Protein Ag1283