missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Epinephrine Hydrochloride, TRC

Product Code. 30134822
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134822 10 g 10g
30132501 25 g 25g
18051340 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134822 Supplier Toronto Research Chemicals Supplier No. TRCE58869810G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-63-5
Molecular Formula C9 H13 N O3 . Cl H
Molecular Weight (g/mol) 219.67
Synonym 1,2-Benzenediol, 4-[1-hydroxy-2-(methylamino)ethyl]-, hydrochloride (1:1), 1,2-Benzenediol, 4-[1-hydroxy-2-(methylamino)ethyl]-, hydrochloride (9CI), 1,2-Benzenediol, 4-[1-hydroxy-2-(methylamino)ethyl]-, hydrochloride, (±)-, Benzyl alcohol, 3,4-dihydroxy-α-[(methylamino)methyl]-, hydrochloride, (±)- (8CI), (±)-Adrenaline hydrochloride, (±)-Epinephrine hydrochloride, Asthmanefrin, Vaponefrin, dl-Adrenaline hydrochloride, dl-Epinephrine hydrochloride, DL-Adrenaline hydrochloride, Racepinephrine hydrochloride, (RS)-4-(1-Hydroxy-2-(methylamino)ethyl)benzene-1,2-diol hydrochloride
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol;hydrochloride
SMILES Cl.CNCC(O)c1ccc(O)c(O)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H13NO3.ClH/c1-10-5-9(13)6-2-3-7(11)8(12)4-6;/h2-4,9-13H,5H2,1H3;1H
Formula Weight 219.0662
Chemical Name or Material DL-Adrenaline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.