missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epiregulin Antibody (6H12), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-21992AF594
This item is not returnable.
View return policy
Description
Epiregulin Monoclonal specifically detects Epiregulin in Mouse samples. It is validated for ELISA.
Specifications
| Epiregulin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| EREG | |
| Mouse recombinant Epiregulin | |
| Primary | |
| Mouse | |
| IgG2 |
| ELISA | |
| 6H12 | |
| ELISA | |
| epiregulin, ER, proepiregulin | |
| Rat | |
| 0.1 mL | |
| 2069 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction