missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epithelia (colonic) Antibody (PR 4D1), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2689DL594
This item is not returnable.
View return policy
Description
Epithelia (colonic) Monoclonal antibody specifically detects Epithelia (colonic) in Human samples. It is validated for Immunohistochemistry
Specifications
| Epithelia (colonic) | |
| Monoclonal | |
| DyLight 594 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry | |
| PR 4D1 | |
| 50mM Sodium Borate | |
| Mucosal scrapings and membrane preparations from normal colorectal epithelium and possibly booster inoculations with the colon carcinoma cell line HT29. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Extracellular Matrix | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction