missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epo Recombinant Rabbit Monoclonal Antibody (HL2752)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556699
This item is not returnable.
View return policy
Description
Epo Recombinant Monoclonal Antibody for Western Blot, Neu
Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.
Specifications
| Epo | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P01588 | |
| EPO | |
| Full length of human EPO. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Neutralization, Western Blot | |
| HL2752 | |
| Unconjugated | |
| EPO | |
| EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2056 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction