missing translation for 'onlineSavingsMsg'
Learn More

Consort™ Epoxy Sealed Standard pH Electrodes

Product Code. 10282875
Change view
Click to view available options
Length (Metric) Cable:
1 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable unitSize
10282875 1 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10282875 Supplier Consort™ Supplier No. SP10T
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Type Standard pH Electrode
For Use With (Application) For pH, conductivity, dissolved oxygen, temperature
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Length (Metric) Cable 1 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.