Learn More
Cytiva Equilibration Tubes
Product Code. 11713773 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
12/Pk.
Unit Size:
12 sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11713773 12/Pk. 12 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11713773 Supplier Cytiva Supplier No. 80646779
Only null left
Add to basket
Add to basket
Includes
Recommendation regarding equilibration solution volumes
Provides a convenient means for equilibrating IPG strips prior to 2-dimension electrophoresis
  • Complete packsize: 12
TRUSTED_SUSTAINABILITY
Specifications
For Use With For equilibrating IPG strips prior to 2-dimension electrophoresis
Quantity 12/Pk.