Learn More
ERAS Monoclonal Antibody (OTI2C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503689S
Description
ERAS Monoclonal antibody specifically detects ERAS in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| ERAS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99439 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1265 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Western Blot | |
| OTI2C6 | |
| Unconjugated | |
| Cnn2 | |
| ecat5; Embryonic stem cell-expressed Ras; ERAS; E-Ras; ES cell expressed Ras; ES cell-expressed Ras; GTPase ERas; Ha-Ras2; HRAS2; HRasp; small GTPase E-Ras; small GTPase protein E-Ras; v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2; v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene | |
| Full length human recombinant protein of human CNN2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.