missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERC1 Mouse anti-Human, Mouse, Rat, Clone: 2A5B9, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | ERC1 |
|---|---|
| Clone | 2A5B9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16888785
|
Proteintech
66681-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16878785
|
Proteintech
66681-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAF-4 is a signal transducer associated with the cytoplasmic domain of the 75 kDa tumor necrosis factor receptor (TNF-R2).Specifications
| ERC1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Cast2, ELKS, ERC 1, ERC1, KIAA1081, Rab6 interacting protein 2, RAB6IP2 | |
| Erc1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2A5B9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat, Human, Mouse | |
| Q811U3, Q8IUD2, Q99MI1 | |
| 111173, 23085, 266806 | |
| ERC1 Fusion Protein Ag17665 | |
| Primary | |
| -20°C | |
| ERC1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title