Learn More
ERCC8 Monoclonal Antibody (OTI9B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808562S
Description
ERCC8 Monoclonal antibody specifically detects ERCC8 in Human samples. It is validated for Western BlotSpecifications
| ERCC8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UHB6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51474 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI9B11 | |
| Unconjugated | |
| LIMA1 | |
| 2410022P04Rik; 2810431L23Rik; 4631412O06Rik; B130065P18Rik; Ckn1; Cockayne syndrome 1 (classical); Cockayne syndrome 1 homolog; cockayne syndrome WD repeat protein CSA; Cockayne syndrome WD repeat protein CSA homolog; Cockayne syndrome WD-repeat protein CSA; CSA; DNA excision repair protein ERCC-8; ERCC excision repair 8, CSA ubiquitin ligase complex subunit; Ercc8; excision repair cross-complementation group 8; excision repair cross-complementing rodent repair deficiency complementation group 8; excision repair cross-complementing rodent repair deficiency, complementation group 8; excision repaiross-complementing rodent repair deficiency, complementation group 8; UVSS2 | |
| Human recombinant protein fragment corresponding to amino acids 1-275 of human LIMA1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.