missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERGIC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16108-1-AP-150UL
This item is not returnable.
View return policy
Description
The MW of this protein is 32 kDa, and this antibody specifically recognizes the 32 kDa protein.
The function of ERGIC1 remains largely unknown. ERGIC1 was highly expressed in most primary prostate tumors. It is an intriguing potential drug target especially for the ERG oncogene expressing tumors.Specifications
| ERGIC1 | |
| Polyclonal | |
| Unconjugated | |
| ERGIC1 | |
| ERGIC 32, ERGIC1, ERGIC32, KIAA1181, NET24 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 287177, 57222, 67458 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q969X5, Q9DC16 | |
| Ergic1 | |
| ERGIC1 Fusion Protein Ag9025 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction