Learn More
Macherey-Nagel™ Ergonomic Crimper
Product Code. 12614577 Shop All Macherey-Nagel Products
Change view
Click to view available options
Material:
Aluminum
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12614577 Aluminum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12614577 Supplier Macherey-Nagel™ Supplier No. 735220
  • ONLINE EXCLUSIVE PRICE
    274.23€
    267.75€ / Pack of 1
    Save 6.48€
    2% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Ergonomic Crimper
For Use With (Equipment) 20 mm Aluminum Crimp Cap
Material Aluminum
No. per Pack 1 Ea.