missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eriodictyol, 94+%

Product Code. 16544474 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16544474 250 mg 250mg
16534474 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16544474 Supplier Thermo Scientific Chemicals Supplier No. 465242500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 552-58-9 | C15H12O6 | 288.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-58-9
Molecular Formula C15H12O6
Molecular Weight (g/mol) 288.26
InChI Key SBHXYTNGIZCORC-ZDUSSCGKSA-N
IUPAC Name (2S)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-1-benzopyran-4-one
SMILES OC1=CC(O)=C2C(=O)C[C@H](OC2=C1)C1=CC=C(O)C(O)=C1

Tekniske data

Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in diluted alkalies
Formula Weight 288.25
Chemical Name or Material Eriodictyol

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.