missing translation for 'onlineSavingsMsg'
Learn More

Eriodictyol, TRC

Product Code. 30385146
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385146 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385146 Supplier Toronto Research Chemicals Supplier No. TRCE6000852.5MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+)-Eriodictyol
CAS 552-58-9
Molecular Formula C15 H12 O6
Quantity 2.5 mg
Synonym 4H-1-Benzopyran-4-one, 2-(3,4-dihydroxyphenyl)-2,3-dihydro-5,7-dihydroxy-, (2S)-, 4H-1-Benzopyran-4-one, 2-(3,4-dihydroxyphenyl)-2,3-dihydro-5,7-dihydroxy-, (S)-, Eriodictyol (6CI), Flavanone, 3',4',5,7-tetrahydroxy- (7CI,8CI), (+)-Eriodictyol, (2S)-2-(3,4-Dihydroxyphenyl)-2,3-dihydro-5,7-dihydroxy-4H-1-benzopyran-4-one, (2S)-Eriodictyol, (S)-3',4',5,7-Tetrahydroxyflavanone, 3',4',5,7-Tetrahydroxyflavanone, Huazhongilexone
InChI Formula InChI=1S/C15H12O6/c16-8-4-11(19)15-12(20)6-13(21-14(15)5-8)7-1-2-9(17)10(18)3-7/h1-5,13,16-19H,6H2/t13-/m0/s1
SMILES Oc1cc(O)c2C(=O)C[C@H](Oc2c1)c3ccc(O)c(O)c3
IUPAC Name (2S)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-2,3-dihydrochromen-4-one
Molecular Weight (g/mol) 288.25
Formula Weight 288.0634
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.