missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK2 Antibody (4C11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60506-0.025ml
This item is not returnable.
View return policy
Description
ERK2 Monoclonal specifically detects ERK2 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ERK2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| MAPK1 | |
| Ni-NTA purified full length recombinant ERK2 expressed in E. Coli strain BL21 (DE3) | |
| Protein A purified | |
| RUO | |
| Primary | |
| ERK2 (4C11C11C4) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot | |
| 4C11 | |
| Unconjugated | |
| EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2 | |
| Mouse | |
| 41 kDa | |
| 0.025 mL | |
| Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Signal Transduction, Tumor Suppressors | |
| 5594 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction