Specifications
| Gene Accession No. | P28482, P28482, P28482, P28482 |
| Preparation Method | This product has been tested by SDS-PAGE and western blot and is suitable for use in Western blot, ELISA, Immunoprecipitation and ChIP. No detection of expected band at ~44 kDa is observed in MAPK1 (ERK2) knockout A549 when compared with unmodified A549 cell lysates by Western blot. |
| Quantity | 100 μg |
| Purity | Multi-step |
| Host Species | Human |
| Content And Storage | Store at -70°C. Avoid freeze/thaw cycles. |
| Research Category | Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Tumor Suppressors |
| Target | ERK2 |
| Synonym | EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2 |
| Gene Symbol | MAPK1 |