missing translation for 'onlineSavingsMsg'
Learn More

Erlose, MP Biomedicals™

Product Code. 11432741 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432741 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11432741 Supplier MP Biomedicals Supplier No. 0219866883
  • ONLINE EXCLUSIVE PRICE
    2360.00€
    2244.90€ / 250mg
    Save 115.10€
    4% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 13101-54-7
Color White
Molecular Formula C18H32O16
MDL Number MFCD00190706
Quantity 250 mg
Synonym erlose, glucosylsucrose, sucrosylglucose, beta-d-fructofuranosyl alpha-d-glucopyranosyl-1->4-alpha-d-glucopyranoside, alpha-maltosyl beta-fructofuranoside, alpha-d-glc-1->4-alpha-d-glc-1->2-beta-d-fru, a-d-glucopyranoside, b-d-fructofuranosyl o-a-d-glucopyranosyl-1, alpha-d-glucopyranoside, beta-d-fructofuranosyl o-alpha-d-glucopyranosyl-1-4, alpha-d-glucopyranosyl-1,2,3,4-alpha-d-glucopyranosyl-1,2-beta-d-fructofuranoside, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-2s,3s,4s,5r-3,4-dihydroxy-2,5-bis hydroxymethyl oxolan-2-yl oxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol
InChI Key FVVCFHXLWDDRHG-KKNDGLDKSA-N
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3(C(C(C(O3)CO)O)O)CO)CO)O)O)O)O
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 504.438
PubChem CID 193471
ChEBI CHEBI:55512
Formula Weight 504.4
Physical Form Crystalline Powder
Chemical Name or Material Erlose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.