Learn More
ERp57/PDIA3 Antibody (CL2446) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36766PCP
Product Code. 30304359
  • 737.00€ / 0.10mL
Description
ERp57/PDIA3 Monoclonal antibody specifically detects ERp57/PDIA3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ERp57/PDIA3
Monoclonal
PerCP
Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3
This antibody was developed using a recombinant protein derived from G5EA52, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers, ER Markers, Membrane Trafficking and Chaperones
2923
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL2446
PBS
Mouse
Protein A purified