missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp72 Mouse anti-Human, Clone: 1D5F3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66365-1-IG-20UL
This item is not returnable.
View return policy
Description
ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.Specifications
| ERp72 | |
| Monoclonal | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13667 | |
| PDIA4 | |
| ERp72 Fusion Protein Ag6873 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 1D5F3 | |
| Unconjugated | |
| PDIA4 | |
| ER protein 70, ER protein 72, ERp 72, ERP70, ERp72, PDIA4, Protein disulfide isomerase A4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 9601 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction