missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ertapenem sodium, 85%

Product Code. 15545296 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545296 250 mg 250mg
15535296 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 15545296 Supplier Thermo Scientific Chemicals Supplier No. 460642500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153773-82-1
Molecular Formula C22H24N3NaO7S
Molecular Weight (g/mol) 497.5
InChI Key ZXNAQFZBWUNWJM-HRXMHBOMSA-M
Synonym 1-azabicyclo 3.2.0 hept-2-ene-2-carboxylic acid, 3-3s,5s-5-3-ca rboxyphenyl amino carbonyl-3-pyrrolidinyl thio-6-1r-1-hydroxyethy l-4-methyl-7-oxo-, monosodium salt, 4r,5s,6s
PubChem CID 131675376
ChEBI CHEBI:60070
IUPAC Name sodium;(2S,4S)-4-[[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl]-N-(3-carboxyphenyl)pyrrolidine-2-carboximidate
SMILES CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=NC4=CC=CC(=C4)C(=O)O)[O-])C(=O)O)C(C)O.[Na+]

Specifications

Melting Point 230.0°C to 234.0°C
Loss on Drying 15% to 19%
Infrared Spectrum Authentic
Assay Percent Range 80% min. (HPLC)
Quantity 250 mg
Beilstein 15,3731
Formula Weight 497.5
Percent Purity 85%
Chemical Name or Material Ertapenem sodium
compliance-icons
Product Identifier
  • Ertapenem sodium
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.