missing translation for 'onlineSavingsMsg'
Learn More

Melford Ertapenem Sodium Salt

Product Code. 30018251
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018251 5 g 5g
30018250 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018251 Supplier Melford Supplier No. E321005.0
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ertapenem Sodium Salt is a potent antibiotic and belongs to the carbapenem class of antibiotics.

Ertapenem Sodium Salt is a potent antibiotic and belongs to the carbapenem class of antibiotics.

It is effective against a broad spectrum of Gram-positive and Gram-negative bacteria, including multidrug-resistant strains.

In the field of microbiology and infectious disease research, Ertapenem Sodium Salt is commonly used as a tool for studying bacterial susceptibility and resistance. It can be employed in antimicrobial susceptibility testing to determine the efficacy of different antibiotics against bacterial isolates.

Specifications

Chemical Name or Material Ertapenem Sodium Salt
CAS 153773-82-1
Quantity 5 g
Molecular Formula C22H24N3NaO7S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.