missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin Stearate, TRC

Product Code. 30012282
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012282 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012282 Supplier Toronto Research Chemicals Supplier No. TRCE6500205G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 643-22-1
Molecular Formula C37 H67 N O13 . C18 H36 O2
Quantity 5 g
Synonym Erythromycin, octadecanoate (1:1), Erythromycin A Stearate, (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-4-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-l-ribo-hexopyranosyl)oxy]-14-ethyl-7,12,13-trihydroxy-3,5,7,9,11,13-hexamethyl-6-[[3,4,6-trideoxy-3-(dimethylamino)-ß-d-xylo-hexopyranosyl]oxy]oxacyclotetradecane-2,10-dione, Erythromycin stearate (6CI,7CI), Erythromycin, octadecanoate (salt) (9CI), Erythromycin, stearate (salt) (8CI), Octadecanoic acid, compd. with erythromycin (1:1) (9CI), Oxacyclotetradecane, erythromycin deriv., Stearic acid, compd. with erythromycin (1:1) (8CI), Abboticine, Bristamycin, Dowmycin E, Eratrex, Erypar, Eryprim, Erythro S, Erythromycin stearic acid salt, Ethril, Ethryn, Gallimycin, Meberyt, OE 7, Pfizer E, SK-Erythromycin, Wemid, Wyamycin S, (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-4-[(2,6-dideoxy-3-C-methyl-3-O-methyl-alpha-l-ribo-hexopyranosyl)oxy]-14-ethyl-7,12,13-trihydroxy-3,5,7,9,11,13-hexamethyl-6-[[3,4,6-trideoxy-3-(dimethylamino)-beta-d-xylo-hexopyranosyl]oxy]oxacyclotetradecane-2,10-dione
InChI Formula InChI=1S/C37H67NO13.C18H36O2/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3;2-17H2,1H3,(H,19,20)/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-;/m1./s1
SMILES CCCCCCCCCCCCCCCCCC(=O)O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-7,12,13-trihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dione;octadecanoic acid
Molecular Weight (g/mol) 1018.4
Formula Weight 1017.7328
Chemical Name or Material Erythromycin A Stearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.