missing translation for 'onlineSavingsMsg'
Learn More

Esomeprazole Sodium, LoGiCal

Product Code. 30457550
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457550 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457550 Supplier LoGiCal Supplier No. LGCFOR3373.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Esomeprazole Sodium
CAS 161796-78-7
Analyte or Component Names Esomeprazole Sodium
Shipping Condition Room Temperature
Molecular Formula C17 H18 N3 O3 S . Na
Certifications/Compliance ISO 9001
Synonym Esomeprazole sodium salt, (S)-(-)-Omeprazole Sodium, (S)-Omeprazole Sodium Salt, Esomeprazole Sodium
InChI Formula InChI=1S/C17H18N3O3S.Na/c1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17;/h5-8H,9H2,1-4H3;/q-1;+1/t24-;/m0./s1
Solution Type Neat
SMILES [Na+].COc1ccc2[n-]c(nc2c1)[S@@](=O)Cc3ncc(C)c(OC)c3C
IUPAC Name sodium;5-methoxy-2-[(S)-(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]benzimidazol-1-ide
Industry Type Clinical Testing
Molecular Weight (g/mol) 367.4
Quantity 10 mg
Formula Weight 367.0967 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.