missing translation for 'onlineSavingsMsg'
Learn More

Esprocarb 10 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30323737
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323737 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323737 Supplier Dr. Ehrenstorfer Supplier No. DRELA13212000AC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Esprocarb
CAS 85785-20-2
Analyte or Component Names Esprocarb
Shipping Condition Room Temperature
Molecular Formula C15 H23 N O S
Concentration 10 μg/mL
Certifications/Compliance ISO/IEC 17025
InChI Formula InChI=1S/C15H23NOS/c1-5-16(13(4)12(2)3)15(17)18-11-14-9-7-6-8-10-14/h6-10,12-13H,5,11H2,1-4H3
Solution Type Single Solution
SMILES CCN(C(C)C(C)C)C(=O)SCc1ccccc1
IUPAC Name S-benzyl N-ethyl-N-(3-methylbutan-2-yl)carbamothioate
Solvent or Matrix Acetone
Industry Type Environmental Testing
Molecular Weight (g/mol) 265.41
Quantity 1 mL
Formula Weight 265.15 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.