Learn More
ESRRB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22644-1-AP-150UL
 View more versions of this product
Product Code. 16802025
  • 410.00€ / 150µL
Description
ESRRB (steroid hormone receptor ERR2) plays a role in maintaice of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survial of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).
Specifications
Specifications
ESRRB
Polyclonal
Unconjugated
ESRRB
DFNB35, ERR2, ERRb, ERRbeta, ERRbeta 2, ESRL2, ESRRB, ESRRB protein, estrogen related receptor beta, NR3B2
Rabbit
Antigen Affinity Chromatography
RUO
2103, 26380, 299210
-20°C
Liquid
Immunoprecipitation, Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95718, P11475, Q61539
ESRRB
ESRRB Fusion Protein Ag18523
150 μL
Primary
Human, Mouse, Rat