missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRRB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22644-1-AP-20UL
This item is not returnable.
View return policy
Description
ESRRB (steroid hormone receptor ERR2) plays a role in maintaice of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survial of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).Specifications
| ESRRB | |
| Polyclonal | |
| Unconjugated | |
| ESRRB | |
| DFNB35, ERR2, ERRb, ERRbeta, ERRbeta 2, ESRL2, ESRRB, ESRRB protein, estrogen related receptor beta, NR3B2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2103, 26380, 299210 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95718, P11475, Q61539 | |
| ESRRB | |
| ESRRB Fusion Protein Ag18523 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction