Learn More
ESRRG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14017-1-AP-20UL
 View more versions of this product
Product Code. 16892474
  • 145.00€ / 20µL
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human ESRRG.
Estrogen-related receptor gamma (ERRG, ERR3, NR3B3) is an orphan member of the nuclear hormone receptor superfamily. ERRG acts as a transcription activator in the absence of a bound ligand. It binds specifically to an estrogen response element and activates reporter genes controlled by estrogen response elements. ERRG also induces the expression of PERM1 in the skeletal muscle.
Specifications
Specifications
ESRRG
Polyclonal
Unconjugated
ESRRG
ERR gamma 2, ERR3, ERRG2, ERRgamma, ESRRG, estrogen related receptor gamma, KIAA0832, NR3B3
Rabbit
Antigen Affinity Chromatography
RUO
2104, 26381, 360896
-20°C
Liquid
Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62508, P62509, P62510
Esrrg
ESRRG Fusion Protein Ag5127
20 μL
Primary
Rat, Mouse, Human