Learn More
ESRRG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14017-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human ESRRG.
Estrogen-related receptor gamma (ERRG, ERR3, NR3B3) is an orphan member of the nuclear hormone receptor superfamily. ERRG acts as a transcription activator in the absence of a bound ligand. It binds specifically to an estrogen response element and activates reporter genes controlled by estrogen response elements. ERRG also induces the expression of PERM1 in the skeletal muscle.Specifications
| ESRRG | |
| Polyclonal | |
| Unconjugated | |
| ESRRG | |
| ERR gamma 2, ERR3, ERRG2, ERRgamma, ESRRG, estrogen related receptor gamma, KIAA0832, NR3B3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2104, 26381, 360896 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62508, P62509, P62510 | |
| Esrrg | |
| ESRRG Fusion Protein Ag5127 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |