Learn More
uvex ESS Hypervision CA FBL Antifog
Product Code. 31751171 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Material Unit Size
31751171 Polycarbonate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31751171 Supplier uvex Supplier No. 9321655
  • ONLINE EXCLUSIVE PRICE
    11.25€
    10.96€ / Pack of 1
    Save 0.29€
    2% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to restore visibility and extend the usable life of safety spectacles through sustainable replacement.
  • Act sustainably by using our replacement lenses to extend the life of your safety spectacles. Scratched lens? No problem-simply replace the lens and continue wearing the same safety spectacles.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Replacement Lens
Disposable Reusable
Lens Material Polycarbonate